MilliporeSigma
  • Diverse reactivity in microwave-promoted catalyst-free coupling of substituted anilines with ethyl trifluoropyruvate and biological evaluation.
Diverse reactivity in microwave-promoted catalyst-free coupling of substituted anilines with ethyl trifluoropyruvate and biological evaluation.
Organic & biomolecular chemistry (2013-07-19)
Chen Zhang, Dao-Min Zhuang, Jia Li, Si-Yuan Chen, Xiao-Long Du, Jian-Yong Wang, Jing-Yun Li, Biao Jiang, Jian-Hua Yao
ABSTRACT
Diverse reactivity by coupling of substituted anilines with ethyl trifluoropyruvate was developed under microwave irradiation without catalysts to generate 3-trifluoromethyl-3-hydroxy oxindoles, aromatic hydroxy trifluoromethyl esters, and 1,2-dicarbonyl compounds in a fast and efficient manner. The plausible mechanism for obtaining different products was proposed. Furthermore, the anti-HIV activity of aromatic hydroxy trifluoromethyl esters was first reported. The best inhibitory activity against wild-type HIV-1 IIIB was exemplified by trifluoromethyloxindole 3q with an IC50 = 5.8 μM, which also displayed potential activity against Y181C mutant virus with an IC50 = 7.5 μM. More significantly, the activities of oxindoles 3q and 3r to inhibit K103N/Y181C double mutant HIV-1 reverse transcriptase (RT) are probably similar to that of the second-generation nonnucleoside inhibitor HBY 097 by docking calculation.
Product Description
Supelco
Aniline, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Pyruvic acid, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Pyruvic acid, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00