MilliporeSigma
Increased expression of c-Jun in nonalcoholic fatty liver disease.
Laboratory investigation; a journal of technical methods and pathology (2014-02-05)
Christoph Dorn, Julia C Engelmann, Michael Saugspier, Andreas Koch, Arndt Hartmann, Martina Müller, Rainer Spang, Anja Bosserhoff, Claus Hellerbrand
ABSTRACT
Overnutrition is the major cause of nonalcoholic fatty liver disease (NAFLD) and its advanced form nonalcoholic steatohepatitis (NASH). We aimed to develop and characterize a murine model, which resembles both the pathology and nutritional situation, of NASH patients in Western societies. Mice were fed with a NASH-inducing diet (ND) containing sucrose, cholesterol and fats rich in saturated fatty acids in a composition, which mimics Western food. After 12 weeks, ND-fed mice revealed obesity and impaired glucose tolerance. In the liver, ND-feeding led to marked steatosis, hepatocellular damage, inflammation and beginning fibrosis. Transcriptome-wide gene expression analysis and search for over-represented transcription factor target sites among the differentially expressed genes identified activator protein-1 (AP-1) as the most likely factor to cause the transcriptional changes in ND livers. Combining differentially expressed gene and protein-protein interaction network analysis identified c-Jun as hub in the largest connected deregulated sub-network in ND livers. Accordingly, ND livers revealed c-Jun-phosphorylation and nuclear translocation. Moreover, hepatic c-Jun expression was enhanced in ND-fed mice. Combined tissue microarray technology and immunohistochemical analysis confirmed enhanced hepatic c-Jun levels in NAFLD patients, which correlated with inflammation, and notably, with the degree of hepatic steatosis. In summary, our new mouse model shows important pathological changes also found in human NASH and indicates c-Jun/AP-1 activation as critical regulator of hepatic alterations. Abundance of c-Jun in NAFLD likely facilitates development and progression of NASH.
Sigma-Aldrich
Palmitic acid, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Anti-CD3, T Cell antibody produced in rabbit, whole antiserum
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, SAJ first grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
Sigma-Aldrich
Sucrose, JIS special grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80