MilliporeSigma
  • Preservation solutions for static cold storage of abdominal allografts: which is best?
Preservation solutions for static cold storage of abdominal allografts: which is best?
Current opinion in organ transplantation (2014-02-21)
Ronald F Parsons, James V Guarrera
ABSTRACT
To update the reader on the recent literature in liver, kidney, pancreas, and intestine static cold preservation, and to identify which solutions are most advantageous for each organ. The comparison of randomized trials of histidine-tryptophan-ketoglutarate (HTK), Celsior, and University of Wisconsin solutions has shown equivalent risk of delayed graft function after kidney transplantation. Similar outcomes have been observed after pancreas preservation with University of Wisconsin, HTK, and Celsior solution. In live-donor liver transplantation, University of Wisconsin and HTK solution have shown equivalent results, whereas in the recent trials of deceased-donor liver transplantation, University of Wisconsin, HTK, and Celsior solutions have shown equivalence. Contrary to the most clinical trials, national registry data in kidney, pancreas, and liver transplantation demonstrate more detrimental effects and earlier graft loss after preservation with HTK versus University of Wisconsin solution. Early outcomes after intestinal transplantation with University of Wisconsin or HTK solution have shown no significant difference and animal studies indicate intraluminal preservation may be beneficial. The University of Wisconsin solution is the standard criterion static cold preservation for the procurement of liver, kidney, pancreas, and intestine. University of Wisconsin, HTK, and Celsior solutions all provide similar allograft outcomes in most clinical trials, but subtle differences have become more apparent in the recent studies and registry reports.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
SAFC
L-Histidine
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Supelco
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard