MilliporeSigma
  • Exercise performance and peripheral vascular insufficiency improve with AMPK activation in high-fat diet-fed mice.
Exercise performance and peripheral vascular insufficiency improve with AMPK activation in high-fat diet-fed mice.
American journal of physiology. Heart and circulatory physiology (2014-02-25)
Kristen A Baltgalvis, Kathy White, Wei Li, Mark D Claypool, Wayne Lang, Raniel Alcantara, Baljit K Singh, Annabelle M Friera, John McLaughlin, Derek Hansen, Kelly McCaughey, Henry Nguyen, Ira J Smith, Guillermo Godinez, Simon J Shaw, Dane Goff, Rajinder Singh, Vadim Markovtsov, Tian-Qiang Sun, Yonchu Jenkins, Gerald Uy, Yingwu Li, Alison Pan, Tarikere Gururaja, David Lau, Gary Park, Yasumichi Hitoshi, Donald G Payan, Todd M Kinsella
ABSTRACT
Intermittent claudication is a form of exercise intolerance characterized by muscle pain during walking in patients with peripheral artery disease (PAD). Endothelial cell and muscle dysfunction are thought to be important contributors to the etiology of this disease, but a lack of preclinical models that incorporate these elements and measure exercise performance as a primary end point has slowed progress in finding new treatment options for these patients. We sought to develop an animal model of peripheral vascular insufficiency in which microvascular dysfunction and exercise intolerance were defining features. We further set out to determine if pharmacological activation of 5'-AMP-activated protein kinase (AMPK) might counteract any of these functional deficits. Mice aged on a high-fat diet demonstrate many functional and molecular characteristics of PAD, including the sequential development of peripheral vascular insufficiency, increased muscle fatigability, and progressive exercise intolerance. These changes occur gradually and are associated with alterations in nitric oxide bioavailability. Treatment of animals with an AMPK activator, R118, increased voluntary wheel running activity, decreased muscle fatigability, and prevented the progressive decrease in treadmill exercise capacity. These functional performance benefits were accompanied by improved mitochondrial function, the normalization of perfusion in exercising muscle, increased nitric oxide bioavailability, and decreased circulating levels of the endogenous endothelial nitric oxide synthase inhibitor asymmetric dimethylarginine. These data suggest that aged, obese mice represent a novel model for studying exercise intolerance associated with peripheral vascular insufficiency, and pharmacological activation of AMPK may be a suitable treatment for intermittent claudication associated with PAD.
Sigma-Aldrich
Cilostazol, ≥98% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
USP
Cilostazol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
SAFC
L-Arginine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Arginine, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00
Sigma-Aldrich
Citrate Synthase Assay Kit, 1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00