MilliporeSigma
  • Rpi-blb2-mediated late blight resistance in Nicotiana benthamiana requires SGT1 and salicylic acid-mediated signaling but not RAR1 or HSP90.
Rpi-blb2-mediated late blight resistance in Nicotiana benthamiana requires SGT1 and salicylic acid-mediated signaling but not RAR1 or HSP90.
FEBS letters (2014-03-04)
Sang-Keun Oh, HyeRan Kim, Doil Choi
ABSTRACT
The Rpi-blb2 recognizes the presence of the Phytophthora infestans AVRblb2 and initiates effector-triggered immunity (ETI). We performed gain-of-function and loss-of-function studies in Nicotiana benthamiana to elucidate Rpi-blb2-mediated resistance to P. infestans. Rpi-blb2 triggered a hypersensitive response through SGT1-mediated, but not RAR-mediated or HSP90-mediated, pathways. NbSGT1 was also required for basal and ETI-mediated by Rpi-blb2 in N. benthamiana. Moreover, salicylic acid (SA) affected basal defense and Rpi-blb2-mediated resistance against P. infestans. The increased susceptibility of Rpi-blb2-transgenic plants in the NahG-background correlated with reduced levels of SA. These findings provide evidence for the roles of SGT1- and SA-signaling in Rpi-blb2-mediated resistance against P. infestans.
Product Description
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization