MilliporeSigma
  • Evaluation of activated sludge for biodegradation of propylene glycol as an aircraft deicing fluid.
Evaluation of activated sludge for biodegradation of propylene glycol as an aircraft deicing fluid.
Water environment research : a research publication of the Water Environment Federation (2014-05-24)
Justin D Delorit, LeeAnn Racz
ABSTRACT
Aircraft deicing fluid used at airport facilities is often collected for treatment or disposal in order to prevent serious ecological threats to nearby surface waters. This study investigated lab scale degradation of propylene glycol, the active ingredient in a common aircraft deicing fluid, by way of a laboratory-scale sequencing batch reactor containing municipal waste water treatment facility activated sludge performing simultaneous organic carbon oxidation and nitrification. The ability of activated sludge to remove propylene glycol was evaluated by studying the biodegradation and sorption characteristics of propylene glycol in an activated sludge medium. The results indicate sorption may play a role in the fate of propylene glycol in AS, and the heterotrophic bacteria readily degrade this compound. Therefore, a field deployable bioreactor may be appropriate for use in flight line applications.
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
(S)-(+)-1,2-Propanediol, 96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
1,2-Propanediol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00