MilliporeSigma
  • Arginine-rich peptides: methods of translocation through biological membranes.
Arginine-rich peptides: methods of translocation through biological membranes.
Current pharmaceutical design (2012-11-13)
Shiroh Futaki, Hisaaki Hirose, Ikuhiko Nakase
ABSTRACT
Intracellular delivery using cell-penetrating peptides (CPPs) has received considerable attention as a promising method for introducing exogenous molecules into cells. The mechanisms that enable efficient internalization of CPPs together with cargo molecules are also of interest. In this review, we describe our current views of membrane translocation of CPPs, especially those rich in arginine, as these peptides represent one of the most frequently employed classes of CPPs.
Product Description
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Arginine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG