MilliporeSigma
  • Role of nasal packing in surgical outcome for chronic rhinosinusitis with polyposis.
Role of nasal packing in surgical outcome for chronic rhinosinusitis with polyposis.
The Laryngoscope (2013-11-29)
Ayşegül Verim, Lütfü Seneldir, Bariş Naiboğlu, Çiğdem Tepe Karaca, Semra Külekçi, Sema Zer Toros, Çağatay Oysu
ABSTRACT
To analyze the differences between biodegradable and nondegradable nasal dressings with regard to their effects on wound healing in the short and medium term and on surgical outcomes in the long term, after endoscopic sinus surgery (ESS). A prospective, randomized, partly blinded, controlled trial. A total of 56 patients undergoing bilateral ESS for chronic rhinosinusitis with polyposis were enrolled and randomized to receive biodegradable (Nasopore; Stryker, Hamilton, ON, Canada) on one side and nondegradable packing (Merocel; Medtronic Xomed, Minneapolis, MN) on the opposite side. Postoperative morbidities (pain, bleeding, facial edema, nasal blockage) related to dressings were assessed on postoperative day 6. Wound healing (edema, crusting, secretions, synechia, granulation tissue formation, and percentage re-epithelialization) were evaluated at 2 weeks, 1 month, 3 months, and 6 months using modified Lund-Kennedy scores. Long-term assessment at 12 months was done using validated Lund-Kennedy scores. Morbidities related to nondegradable packing were significantly higher than with degradable packing (Wilcoxon signed rank test, P<.01). Pain, bleeding, nasal blockage, and facial edema were significantly less with absorbable packing. No statistically significant difference was found between sinonasal cavities packed with biodegradable or nondegradable materials with regard to healing scores and percentage of re-epithelialization at 2 weeks, 1 month, 3 months, 6 months, and surgical outcomes at 1 year (P>.05). However, healing at 6 months was correlated with the preoperative Lund-Mackay radiology and surgery scores (Spearman's rho correlation test, P<.05 and P<.01, respectively). No significant healing or surgical outcome differences were found between biodegradable and nondegradable packing. However pain, bleeding, nasal blockage, and facial edema were lower with biodegradable packing. 1b.
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sign Into View Organizational & Contract Pricing
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), SAJ first grade, degree of polymerization 500
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, 10%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sign Into View Organizational & Contract Pricing