MilliporeSigma
  • Estrogen regulation of the brain renin-angiotensin system in protection against angiotensin II-induced sensitization of hypertension.
Estrogen regulation of the brain renin-angiotensin system in protection against angiotensin II-induced sensitization of hypertension.
American journal of physiology. Heart and circulatory physiology (2014-05-27)
Baojian Xue, Zhongming Zhang, Terry G Beltz, Fang Guo, Meredith Hay, Alan Kim Johnson
ABSTRACT
This study investigated sex differences in the sensitization of angiotensin (ANG) II-induced hypertension and the role of central estrogen and ANG-(1-7) in this process. Male and female rats were implanted for telemetered blood pressure (BP) recording. A subcutaneous subpressor dose of ANG II was given alone or concurrently with intracerebroventricular estrogen, ANG-(1-7), an ANG-(1-7) receptor antagonist A-779 or vehicle for 1 wk (induction). After a 1-wk rest (delay), a pressor dose of ANG II was given for 2 wk (expression). In males and ovariectomized females, subpressor ANG II had no sustained effect on BP during induction, but produced an enhanced hypertensive response to the subsequent pressor dose of ANG II during expression. Central administration of estrogen or ANG-(1-7) during induction blocked ANG II-induced sensitization. In intact females, subpressor ANG II treatment produced a decrease in BP during induction and delay, and subsequent pressor ANG II treatment given during expression produced only a slight but significant increase in BP. However, central blockade of ANG-(1-7) by intracerebroventricular infusion of A-779 during induction restored the decreased BP observed in females during induction and enhanced the pressor response to the ANG II treatment during expression. RT-PCR analyses indicated that estrogen given during induction upregulated mRNA expression of the renin-angiotensin system (RAS) antihypertensive components, whereas both central estrogen and ANG-(1-7) downregulated mRNA expression of RAS hypertensive components in the lamina terminalis. The results indicate that females are protected from ANG II-induced sensitization through central estrogen and its regulation of brain RAS.
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®