MilliporeSigma
  • Advantage of lutetium-177 versus radioiodine immunoconjugate in targeted radionuclide therapy of b-cell tumors.
Advantage of lutetium-177 versus radioiodine immunoconjugate in targeted radionuclide therapy of b-cell tumors.
Anticancer research (2014-07-02)
Ada Repetto-Llamazares, Nasir Abbas, Øyvind S Bruland, Jostein Dahle, Roy H Larsen
ABSTRACT
We herein report a comparison of the radiolabels 177Lu and 125I bound to the monoclonal antibody HH1 that targets the CD37 antigen expressed on non-Hodgkin B-cell lymphomas. Mixtures of 177Lu and 125I-labeled HH1 antibody were co-injected into nude mice carrying Ramos xenografts and the biodistribution using the paired label format allowing tracer comparisons in each individual mouse. Products of the two radionuclides had very similar immunoractivity in vitro but showed different properties in vivo. Both products had relevant stability in blood and most normal tissues in nude mice carrying subcutaneous Ramos xenografts. However, both the tumor uptake and retention were significantly higher for 177Lu vs. 125I labeled HH1. The tumor to normal tissue ratios were several-fold improved for 177Lu compared to radioiodine labeled antibodies. The data presented herein support the evaluation of CD37 as a target for clinical 177Lu-based radioimmunotherapy against b-cell malignancies.
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)
Lutetium, powder, 1g, max. particle size 500 micron, 99.9%
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%