USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Reagent Alcohol, reagent grade
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chlorhexidine, ≥99.5%
Sign Into View Organizational & Contract Pricing Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sign Into View Organizational & Contract Pricing Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing