MilliporeSigma
  • Time course of asymmetric dimethylarginine and symmetric dimethylarginine levels after successful renal transplantation.
Time course of asymmetric dimethylarginine and symmetric dimethylarginine levels after successful renal transplantation.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2014-06-25)
Kathleen J Claes, Bert Bammens, Dirk R Kuypers, Björn Meijers, Maarten Naesens, Ben Sprangers, Yves Vanrenterghem, Evelyne Lerut, Henriette de Loor, Pieter Evenepoel
ABSTRACT
Although renal transplantation (Tx) improves the outcome of patients with renal disease, cardiovascular (CV) risk remains high. Recently, it was demonstrated that asymmetric dimethylarginine (ADMA) levels predict CV events and graft survival in renal transplant recipients (RTRs). Little is known about the impact of renal Tx on the plasma levels of ADMA and symmetric dimethylarginine (SDMA). The present study aimed to define the time course of ADMA and SDMA after Tx. We prospectively followed 167 incident RTRs with visits at the time of Tx and 3 and 12 months thereafter. At all visits, demographics and relevant biochemistry were recorded and blood was sampled for analysis of ADMA and SDMA (high-performance liquid chromatography). Eighty-four patients had an additional sampling in the immediate postoperative period. In a case-controlled substudy (n = 31), we compared ADMA and SDMA levels between RTRs and chronic kidney disease (CKD) patients, matched for glomerular filtration rate, gender, age, CV history and diabetes. Overall, plasma ADMA and SDMA levels decreased after Tx. The decline of SDMA was more pronounced and paralleled the recovery of renal function. Interestingly, the decline of ADMA was preceded by an increase in the immediate postoperative period. In the case-controlled substudy, SDMA levels were similar, whereas ADMA levels were significantly higher in RTRs compared with the CKD counterparts (P = 0.003). ADMA levels follow a biphasic pattern after successful renal Tx with a transient rise in the immediate postoperative period followed by a decline. Levels remain elevated compared with CKD patients, matched for age, gender, diabetes, CV history and renal function.
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
SAFC
L-Arginine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Arginine, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 22, 2025
$131.00
1 kg
Estimated to ship on March 22, 2025
$328.00
50 kg
Estimated to ship on March 22, 2025
$12,800.00