MilliporeSigma
  • Influence of l-carnosine on pro-antioxidant status in elite kayakers and canoeists.
Influence of l-carnosine on pro-antioxidant status in elite kayakers and canoeists.
Acta physiologica Hungarica (2014-09-10)
Malgorzata Slowinska-Lisowska, A Zembron-Lacny, M Rynkiewicz, T Rynkiewicz, W Kopec
ABSTRACT
Carnosine is a dipeptide formed from the amino acids β-alanine and histidine and found in large amounts in the brain and muscle, especially fast twitch muscle. Carnosine has an antioxidant role and accounts for about 10% of the muscle's ability to buffer the H+ ions produced by high intensity exercise. Due to the interesting role of carnosine, the aim of the study was observe the effects of carnosine intake on pro-antioxidant status in highly trained athletes exposed to intense exercise.Fourteen male athletes from the Polish national kayak and canoe teams participated in placebo-controlled and cross-over study. The athletes were supplemented with 4 g/d carnosine for 14 days. Blood samples were collected before and 30 min, 24 h and 48 h after 2000 m exercise trial. In blood, hydrogen peroxide (H2O2), nitric oxide (NO), markers of RO/NS activity 8-isoprostanes and 3-nitrotyrosine, total (GSHt) and oxidised glutathione (GSSG), antioxidant status (APO) and superoxide dismutase (SOD) were determined. There were not observed statistically significant differences in exercise-induced changes in H2O2 and NO concentrations and SOD activity after carnosine intake. However, carnosine prevented an increase in 8-isoprostanes, 3-nitrotyrosine and GSSG concentrations as well as elevated redox status (GSHt-2GSSG)/GSSG at post-exercise period.Although, oral supplementation with 4 g carnosine did not affect RO/NS generation, it significantly attenuated exercise-induced glutathione loss, reduced oxidation/nitration markers concentration and SOD activity. These results suggest that carnosine could provide antioxidative protection for highly trained athletes.
Product Description
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Lactic acid, JIS special grade, 85.0-92.0%
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
L-Carnosine, ~99%, crystalline
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Tyrosine
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactic acid, natural, ≥85%
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material