MilliporeSigma
  • Spatiotemporal distribution and mass loadings of perfluoroalkyl substances in the Yangtze River of China.
Spatiotemporal distribution and mass loadings of perfluoroalkyl substances in the Yangtze River of China.
The Science of the total environment (2014-07-02)
Chang-Gui Pan, Guang-Guo Ying, Jian-Liang Zhao, You-Sheng Liu, Yu-Xia Jiang, Qian-Qian Zhang
ABSTRACT
A systematic investigation into contamination profiles of eighteen perfluoroalkyl substances (PFASs) in both surface water and sediments of Yangtze River was carried out by using high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) in summer and winter of 2013. The total concentrations of the PFASs in the water and sediment of Yangtze River ranged from 2.2 to 74.56 ng/L and 0.05 to 1.44 ng/g dry weights (dw), respectively. The PFAS concentrations were correlated to some selected water quality parameters such as pH, total phosphorus (TP), total nitrogen (TN) and conductivity in water, and some sediment properties, such as total organic carbon (TOC), TP, and TN in sediment. The monitoring results for the water and sediment samples showed no obvious seasonal variations. Among the selected 18 PFASs, perfluorooctanoic acid (PFOA) was the dominant PFAS compound found both in water and sediment for the two seasons with its maximum concentration of 18.03 ng/L in water and 0.72 ng/g in sediment, followed by perfluorobutane sulfonic acid (PFBS) with its maximum concentration of 41.9 ng/L in water in Wuhan, whereas the lowest concentrations of PFASs were observed at Poyang lake. The annual loadings of PFOA, perfluorohexanoic acid (PFHxA), PFBS, perfluorooctane sulfonic acid (PFOS) and the total PFASs in the Yangtze River were 6.8 tons, 2.2 tons, 8.2 tons, 0.88 tons, and 20.7 tons, respectively. Wuhan and Er'zhou of Hubei contributed the most amounts of PFASs into the Yangtze River. A correlation was found between some PFASs, for example PFBS and PFOS, which suggests that both of these PFASs originate from common sources in the region.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles