MilliporeSigma
  • 5-Aminosalicylic acid attenuates allergen-induced airway inflammation and oxidative stress in asthma.
5-Aminosalicylic acid attenuates allergen-induced airway inflammation and oxidative stress in asthma.
Pulmonary pharmacology & therapeutics (2014-08-08)
K Rama Satyanarayana Raju, M N Sathish Kumar, Saurabh Gupta, Srinivas T Naga, Jaya K Shankar, Vishakantha Murthy, Subba Rao V Madhunapanthula, Shashank Mulukutla, Nilesh S Ambhore, Shashank Tummala, V J Vishnuvarthan, Afzal Azam, Kannan Elango
ABSTRACT
Pro-inflammatory cytokines regulate the magnitude of allergic reactions during asthma. Tumor necrosis factor--alpha (TNF-α), interleukin-6 (IL-6) and interleukin-13 (IL-13) play a crucial role in aggravating the inflammatory conditions during allergic asthma. In addition, oxidative stress contributes to the pathogenesis of asthma by altering the physiological condition resulting in the development of status asthmaticus. Anti-inflammatory corticosteroids are being widely used for treating allergic asthma. In the present study 5-aminosalicylic acid (5-ASA), a salicylic acid derivative, was evaluated, in vivo for its potential to suppress TNF-α, IL-6 and IL-13 using ovalbumin (OVA) induced allergic asthma in Balb/C mice. Oral administration of 65, 130 and 195 mg/kg 5-ASA significantly reduced the OVA induced total and differential leucocyte count, TNF-α, IL-6, IL-13, nitrite, nitrate, MDA, MPO and TPL levels in the lung lavage samples. Collectively, these findings suggest that 5-ASA is a potent immunomodulator and suppresses key Th2 cytokines production and oxidative stress in OVA-induced asthma.
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium bromide, 99.999% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ammonium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Ammonium bromide, ACS reagent, ≥99.0%
Mesalazine, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material