MilliporeSigma
  • Hydrophilic interaction liquid chromatography as second dimension in multidimensional chromatography with an anionic trapping strategy: application to prostate-specific antigen quantification.
Hydrophilic interaction liquid chromatography as second dimension in multidimensional chromatography with an anionic trapping strategy: application to prostate-specific antigen quantification.
Journal of chromatography. A (2014-06-17)
Romain Simon, Sébastien Passeron, Jérôme Lemoine, Arnaud Salvador
ABSTRACT
Liquid chromatography (LC) coupled with tandem mass spectrometry (MS-MS) in selected reaction monitoring mode (SRM) has become a widely used technique for the quantification of protein biomarkers in plasma and has already proven to give similar results compared to the conventional immunoassays. To improve the lack of insufficient sensitivity for quantification of low abundance protein, we propose a new two dimensional liquid chromatography (2D-LC-SRM) method for the quantitation of prostate specific antigen (PSA) in human plasma. The method centers on anion exchange cartridge between reversed-phase chromatography and hydrophilic interaction liquid chromatography (HILIC) in an on-line arrangement. The use of the anionic cartridge allows an easier online transfer of the analytes between both dimensions. Moreover, it provides an additional selectivity since the more basic peptides are not retained on this support. This setup has been applied to the quantification of prostate specific antigen (PSA) protein in plasma on a previous generation of mass spectrometer, which enabled a limit of quantification (LOQ) of 1ng/mL without any upfront immuno-depletion or intense off-line fractionation before the SRM analysis. The obtained LOQ is compatible with the required sensitivity for the clinically relevant plasma-based PSA tests.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)