MilliporeSigma
  • Glutathione reductase activity with an oxidized methylated glutathione analog.
Glutathione reductase activity with an oxidized methylated glutathione analog.
Journal of enzyme inhibition and medicinal chemistry (2013-07-03)
Brant L Kedrowski, Jonathan H Gutow, Gorman Stock, Maureen Smith, Chondrea Jordan, Douglas S Masterson
ABSTRACT
The activity of glutathione reductase with an unnatural analog of oxidized glutathione was explored. The analog, L-γ-glutamyl-2-methyl-L-cysteinyl-glycine disulfide, places an additional methyl group on the alpha position of each of the central cysteine residues, which significantly increases steric bulk near the disulfide bond. Glutathione reductase was completely unable to catalyze the sulfur-sulfur bond reduction of the analog. Additionally, enzyme kinetics experiments indicated that the analog acts as a competitive inhibitor of glutathione reductase. Computational studies confirm that the methylated analog fits within the active site of the enzyme but its disulphide bond geometry is altered, preventing reduction by the enzyme. The substitution of (R)-2-methylcysteine in place of natural (R)-cysteine in peptides constitutes a new strategy for stabilizing disulphide bonds from enzyme-catalyzed degradation.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard