MilliporeSigma
  • Probiotic Lactobacillus rhamnosus GG (LGG) and prebiotic prevent neonatal inflammation-induced visceral hypersensitivity in adult rats.
Probiotic Lactobacillus rhamnosus GG (LGG) and prebiotic prevent neonatal inflammation-induced visceral hypersensitivity in adult rats.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2014-10-10)
P Kannampalli, S Pochiraju, M Chichlowski, B M Berg, C Rudolph, M Bruckert, A Miranda, J N Sengupta
ABSTRACT
Increasing evidence indicates a positive effect of probiotics on the nervous system. The objective of this study was to determine if probiotic Lactobacillus rhamnosus GG (LGG) and/or prebiotics polydextrose/galactooligosaccharide (PDX/GOS) can alter the colonic sensitivity in a neonatal rat model of chronic visceral hyperalgesia and to determine whether altered sensitivity is associated with changes in neurotransmitter levels in the brain. Chronic visceral hyperalgesia was induced in rats by intracolonic administration of zymosan for 3 days during postnatal day 14-16 (P14-P16). After weaning (P21), these pups were divided into groups that received either (1) control diet (CD), (2) PDX/GOS, (3) LGG, or (4) PDX/GOS + LGG. These diets were continued until visceral sensitivity was tested at P60. The viscero-motor response (VMR) to graded colorectal distension (CRD) was determined by measuring the electromyographic (EMG) activity from the abdominal external oblique muscles. The levels of neurotransmitters and biogenic amines were quantified in the frontal cortex, subcortex, brain stem, and cerebellum. At P60, the VMR to CRD in the neonatal zymosan-treated rats was significantly higher than neonatal saline-treated rats. In contrast, neonatal zymosan-treated rats that received PDX/GOS or LGG did not exhibit visceral hyperalgesia. The levels of serotonin, noradrenaline, and dopamine were significantly altered in LGG-treated rats compared to other groups. Results document that in rats LGG can attenuate neonatally induced chronic visceral pain measured in adulthood. Prolonged intake of LGG alters some key brain neurotransmitters and biogenic amines that could be involved in pain modulation.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetonitrile, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Supelco
Carprofen, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetonitrile, HPLC grade, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, for chromatography
SKU
Pack Size
Availability
Price
Quantity
Carprofen, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Carprofen, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O