MilliporeSigma
  • A validated dispersive liquid-liquid microextraction method for extraction of ochratoxin A from raisin samples.
A validated dispersive liquid-liquid microextraction method for extraction of ochratoxin A from raisin samples.
Journal of food science and technology (2015-04-02)
Rouhollah Karami-Osboo, Ramin Miri, Katayoun Javidnia, Farzad Kobarfard, Mohammad Hossein Shojaee AliAbadi, Mehdi Maham
ABSTRACT
A method based on dispersive liquid-liquid microextraction (DLLME) was developed for the quantitative extraction of Ochratoxin A (OTA) from raisin samples. The influence of various parameters on the recovery of OTA such as type and volume of DLLME extractant, centrifuging and sonication time, also volume of deionized water was investigated. Recovery values under the optimum conditions were between 68.6 and 85.2 %, the inner and intra-day precision expressed as relative standard deviation (RSD%, n = 3), were less than 15 % at spiking levels of 2.5-30 μg kg(-1). Linearity was studied from 0.5 to 30 μg L(-1), and the limits of detection (LOD) and quantification (LOQ) were 0.7 and 2.0 μg kg(-1), respectively. Real samples were analyzed by DLLME method and compared with confirmative immunoaffinity Column Chromatography (IAC) clean-up. Low cost, simplicity of operation, speed and minimum consumption of organic solvent were the main advantages of proposed method. The mean contamination of samples was 0.88 μg kg(-1) that was lower than European Legal Limit.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%