MilliporeSigma
  • Alleviation of hepatic injury by chrysin in cisplatin administered rats: probable role of oxidative and inflammatory markers.
Alleviation of hepatic injury by chrysin in cisplatin administered rats: probable role of oxidative and inflammatory markers.
Pharmacological reports : PR (2014-12-03)
Muneeb U Rehman, Nemat Ali, Summya Rashid, Tyan Jain, Sana Nafees, Mir Tahir, Abdul Quaiyoom Khan, Abdul Lateef, Rehan Khan, Oday O Hamiza, Syed Kazim, Wajhul Qamar, Sarwat Sultana
ABSTRACT
Cisplatin is an effective and extensively used chemotherapeutic agent to treat range of malignancies, but its therapeutic use is limited because of dose-dependent nephrotoxicity and hepatotoxicity. Several published reports advocate that supplementation with antioxidant can influence cisplatin induced hepatic damage. In the present study the Wistar rats were subjected to concurrent prophylactic oral treatment of chrysin (25 and 50mg/kgb.wt.) against the hepatotoxicity induced by intraperitoneal administration of cisplatin (7.5mg/kgb.wt.). Efficacy of chrysin against the hepatotoxicity was evaluated in terms of biochemical estimation of antioxidant enzyme activities, histopathological changes and expression levels of molecular markers of inflammation. Chrysin ameliorated cisplatin-induced lipid peroxidation, xanthine oxidase activity, glutathione depletion, decrease in antioxidant (catalase, glutathione reductase, superoxide dismutase, glutathione peroxidase and glucose-6 phosphate dehydrogenase) and phase-II detoxifying (glutathione-S-transferase and quinone reductase) enzyme activities. Chrysin also attenuated expression of COX-2, iNOS and levels of NFκB and TNF-α, and hepatic tissue damage which were induced by cisplatin. Histological findings further supported the protective effects of chrysin against cisplatin-induced hepatic damage. The results of the present study demonstrate that oxidative stress and inflammation are closely associated with cisplatin-induced toxicity and chrysin shows the protective efficacy against cisplatin-induced hepatotoxicity possibly via attenuating the oxidative stress and inflammatory response.
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Perchloric acid, SAJ super special grade, 60.0-62.0%
Sigma-Aldrich
Perchloric acid, 0.1 M
Supelco
Chrysin, analytical standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%