MilliporeSigma
  • The tyrosine phosphatase PTPN14 (Pez) inhibits metastasis by altering protein trafficking.
The tyrosine phosphatase PTPN14 (Pez) inhibits metastasis by altering protein trafficking.
Science signaling (2015-02-19)
Leila Belle, Naveid Ali, Ana Lonic, Xiaochun Li, James L Paltridge, Suraya Roslan, David Herrmann, James R W Conway, Freya K Gehling, Andrew G Bert, Lesley A Crocker, Anna Tsykin, Gelareh Farshid, Gregory J Goodall, Paul Timpson, Roger J Daly, Yeesim Khew-Goodall
ABSTRACT
Factors secreted by tumor cells shape the local microenvironment to promote invasion and metastasis, as well as condition the premetastatic niche to enable secondary-site colonization and growth. In addition to this secretome, tumor cells have increased abundance of growth-promoting receptors at the cell surface. We found that the tyrosine phosphatase PTPN14 (also called Pez, which is mutated in various cancers) suppressed metastasis by reducing intracellular protein trafficking through the secretory pathway. Knocking down PTPN14 in tumor cells or injecting the peritoneum of mice with conditioned medium from PTPN14-deficient cell cultures promoted the growth and metastasis of breast cancer xenografts. Loss of catalytically functional PTPN14 increased the secretion of growth factors and cytokines, such as IL-8 (interleukin-8), and increased the abundance of EGFR (epidermal growth factor receptor) at the cell surface of breast cancer cells and of FLT4 (vascular endothelial growth factor receptor 3) at the cell surface of primary lymphatic endothelial cells. We identified RIN1 (Ras and Rab interactor 1) and PRKCD (protein kinase C-δ) as binding partners and substrates of PTPN14. Similar to cells overexpressing PTPN14, receptor trafficking to the cell surface was inhibited in cells that lacked PRKCD or RIN1 or expressed a nonphosphorylatable RIN1 mutant, and cytokine secretion was decreased in cells treated with PRKCD inhibitors. Invasive breast cancer tissue had decreased expression of PTPN14, and patient survival was worse when tumors had increased expression of the genes encoding RIN1 or PRKCD. Thus, PTPN14 prevents metastasis by restricting the trafficking of both soluble and membrane-bound proteins.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
O-Phospho-L-tyrosine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, tablet
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, 1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
SAFC
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on January 23, 2025
Details...
₩430,259