MilliporeSigma
  • Development and validation of a sensitive method for simultaneous determination of eight β₂-agonists in pork by ultrasonic-assisted extraction and liquid chromatography/tandem mass spectrometry.
Development and validation of a sensitive method for simultaneous determination of eight β₂-agonists in pork by ultrasonic-assisted extraction and liquid chromatography/tandem mass spectrometry.
Journal of chromatographic science (2014-04-29)
Shengdong Pan, Lixin Zhou, Yonggang Zhao, Xiaohong Chen, Xiaoping Li, Micong Jin
ABSTRACT
A simple and sensitive ultrasonic-assisted extraction coupled with liquid chromatography-tandem quadrupole mass spectrometry was developed for the simultaneous determination of eight β2-agonists (i.e., phenylethanolamine, fenoterol, formoterol, clenbuterol, ractopamine, salbutamol, terbutaline and tulobuterol) in pork. The recovery rate of ultrasonic-assisted extraction was compared with that of enzymolysis. Results showed that ultrasonic-assisted extraction is a satisfactory method for the treatment of pork samples, yielding β2-agonist recovery rates of 82.0-114.0%. In contrast, most of the recovery rates obtained by enzymolysis were <80%. Linearities obtained ranged from 0.1 to 16.0 μg kg(-1) for terbutaline, formoterol, tulobuterol and phenylethanolamine, 0.2 to 16.0 μg kg(-1) for salbutamol, fenoterol and ractopamine, and 0.2 to 40.0 μg kg(-1) for clenbuterol. The correlation coefficients were >0.9945 for all analytes. The lower limits of quantification were between 0.086 and 0.20 μg kg(-1). The applicability of the proposed method for detecting and quantifying β2-agonists was demonstrated in the analysis of 50 pork samples.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v)
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%