MilliporeSigma
  • Speed control for neuronal migration in the postnatal brain by Gmip-mediated local inactivation of RhoA.
Speed control for neuronal migration in the postnatal brain by Gmip-mediated local inactivation of RhoA.
Nature communications (2014-07-31)
Haruko Ota, Takao Hikita, Masato Sawada, Tomoki Nishioka, Mami Matsumoto, Masayuki Komura, Akihisa Ohno, Yukiyo Kamiya, Takuya Miyamoto, Naoya Asai, Atsushi Enomoto, Masahide Takahashi, Kozo Kaibuchi, Kazuya Sobue, Kazunobu Sawamoto
ABSTRACT
Throughout life, new neurons generated in the ventricular-subventricular zone take the long journey to the olfactory bulb. The intracellular mechanisms that precisely control the neurons' migration speed, enabling their well-organized movement, remain unclear. Rho signalling is known to affect the morphology and movement of various cell types, including neurons. Here we identify Gem-interacting protein (Gmip), a RhoA-specific GTPase-activating protein, as a key factor in saltatory neuronal migration. RhoA is activated at the proximal leading process of migrating neurons, where Gmip is also localized and negatively regulates RhoA. Gmip controls the saltatory movement of neurons that regulate their migration speed and 'stop' positions in the olfactory bulb, thereby altering the neural circuitry. This study demonstrates that Gmip serves as a brake for the RhoA-mediated movement of neuronal somata, and highlights the significance of speed control in the well-organized neuronal migration and the maintenance of neuronal circuits in the postnatal brain.
Product Description
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard