MilliporeSigma
  • Palmitoylation of the Alphacoronavirus TGEV spike protein S is essential for incorporation into virus-like particles but dispensable for S-M interaction.
Palmitoylation of the Alphacoronavirus TGEV spike protein S is essential for incorporation into virus-like particles but dispensable for S-M interaction.
Virology (2014-08-13)
Sandra Gelhaus, Bastian Thaa, Kathrin Eschke, Michael Veit, Christel Schwegmann-Weßels
ABSTRACT
The spike protein S of coronaviruses contains a highly conserved cytoplasmic cysteine-rich motif adjacent to the transmembrane region. This motif is palmitoylated in the Betacoronaviruses MHV and SARS-CoV. Here, we demonstrate by metabolic labeling with [(3)H]-palmitic acid that the S protein of transmissible gastroenteritis coronavirus (TGEV), an Alphacoronavirus, is palmitoylated as well. This is relevant for TGEV replication as virus growth was compromised by the general palmitoylation inhibitor 2-bromopalmitate. Mutation of individual cysteine clusters in the cysteine-rich motif of S revealed that all cysteines must be replaced to abolish acylation and incorporation of S into virus-like particles (VLP). Conversely, the interaction of S with the M protein, essential for VLP incorporation of S, was not impaired by lack of palmitoylation. Thus, palmitoylation of the S protein of Alphacoronaviruses is dispensable for S-M interaction, but required for the generation of progeny virions.
Sigma-Aldrich
Sucrose, JIS special grade
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
SAFC
L-Glutamine
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, SAJ first grade
Sucrose, European Pharmacopoeia (EP) Reference Standard