MilliporeSigma
SH2B3 Is a Genetic Determinant of Cardiac Inflammation and Fibrosis.
Circulation. Cardiovascular genetics (2015-01-30)
Michael J Flister, Matthew J Hoffman, Angela Lemke, Sasha Z Prisco, Nathan Rudemiller, Caitlin C O'Meara, Shirng-Wern Tsaih, Carol Moreno, Aron M Geurts, Jozef Lazar, Neeta Adhikari, Jennifer L Hall, Howard J Jacob
ABSTRACT
Genome-wide association studies are powerful tools for nominating pathogenic variants, but offer little insight as to how candidate genes affect disease outcome. Such is the case for SH2B adaptor protein 3 (SH2B3), which is a negative regulator of multiple cytokine signaling pathways and is associated with increased risk of myocardial infarction (MI), but its role in post-MI inflammation and fibrosis is completely unknown. Using an experimental model of MI (left anterior descending artery occlusion/reperfusion injury) in wild-type and Sh2b3 knockout rats (Sh2b3(em2Mcwi)), we assessed the role of Sh2b3 in post-MI fibrosis, leukocyte infiltration, angiogenesis, left ventricle contractility, and inflammatory gene expression. Compared with wild-type, Sh2b3(em2Mcwi) rats had significantly increased fibrosis (2.2-fold; P<0.05) and elevated leukocyte infiltration (>2-fold; P<0.05), which coincided with decreased left ventricle fractional shortening (-Δ11%; P<0.05) at 7 days post left anterior descending artery occlusion/reperfusion injury. Despite an increased angiogenic potential in Sh2b3(em2Mcwi) rats (1.7-fold; P<0.05), we observed no significant differences in left ventricle capillary density between wild-type and Sh2b3(em2Mcwi) rats. In total, 12 genes were significantly elevated in the post left anterior descending artery occluded/reperfused hearts of Sh2b3(em2Mcwi) rats relative to wild-type, of which 3 (NLRP12, CCR2, and IFNγ) were significantly elevated in the left ventricle of heart failure patients carrying the MI-associated rs3184504 [T] SH2B3 risk allele. These data demonstrate for the first time that SH2B3 is a crucial mediator of post-MI inflammation and fibrosis.
Product Description
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Supelco
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
USP
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard