MilliporeSigma
  • Total synthesis of putative montamine and a proposed structural reassignment.
Total synthesis of putative montamine and a proposed structural reassignment.
Organic & biomolecular chemistry (2014-07-24)
Lachlan M Blair, Elizabeth A Colby Davie, Jonathan Sperry
ABSTRACT
The natural product montamine was originally assigned as a homodimer of moschamine linked by a N-N' bond at the serotonin side-chain. A total synthesis of the reported structure has shown this to be incorrect. Analysis of the spectroscopic data suggests that the dimerization site has been incorrectly assigned, and montamine is likely to be a 4,4'-bismoschamine natural product previously described in the literature.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Cesium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Cesium carbonate, 99.95% trace metals basis
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Cesium carbonate, puriss. p.a., ≥99.0%
Sigma-Aldrich
Cesium carbonate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%