MilliporeSigma
  • Cardiolipin-incorporated liposomes with surface CRM197 for enhancing neuronal survival against neurotoxicity.
Cardiolipin-incorporated liposomes with surface CRM197 for enhancing neuronal survival against neurotoxicity.
International journal of pharmaceutics (2014-07-08)
Yung-Chih Kuo, Yu-Chuan Liu
ABSTRACT
CRM197-grafted liposomes containing cardiolipin (CL) (CRM197/CL-liposomes) were used to enhance the permeability of neuron growth factor (NGF) across the blood-brain barrier (BBB) for promoting the neuroprotective effect of NGF. CRM197/CL-liposoms were incubated with a monolayer of human astrocyte (HA)-regulated human brain-microvascular endothelial cells (HBMECs) and employed to rescue SK-N-MC cells with insult of fibrillar β-amyloid peptide (1-42) (Aβ1-42). An increase in the CL mole percentage enhanced the particle size, absolute value of zeta potential, NGF entrapment efficiency, CRM197 grafting efficiency, viability of HBMECs, HAs, and SK-N-MC cells, and BBB permeability of propidium iodide (PI) and NGF, and reduced the transendothelial electrical resistance (TEER). In addition, an increase in the CRM197 weight percentage increased the particle size, absolute value of zeta potential, viability of HBMECs and HAs, and BBB permeability of PI and NGF, and decreased the CRM197 grafting efficiency and TEER. CRM197/CL-liposomes have the ability to target the BBB and to reduce neurotoxicity of Aβ142 and can be promising formulations for treating Alzheimer's disease in future medicinal application.
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%