Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Supelco
Hexadecane, analytical standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Imidazole, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sign Into View Organizational & Contract Pricing Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri™, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri™
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri™, free-flowing
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99%
Sign Into View Organizational & Contract Pricing USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sign Into View Organizational & Contract Pricing Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sign Into View Organizational & Contract Pricing USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Supelco
Pentane, analytical standard
Sign Into View Organizational & Contract Pricing