MilliporeSigma
  • Dapper-1 is essential for Wnt5a induced cardiomyocyte hypertrophy by regulating the Wnt/PCP pathway.
Dapper-1 is essential for Wnt5a induced cardiomyocyte hypertrophy by regulating the Wnt/PCP pathway.
FEBS letters (2014-06-01)
Marco Hagenmueller, Johannes H Riffel, Elmar Bernhold, Jingjing Fan, Hugo A Katus, Stefan E Hardt
ABSTRACT
The Wnt signaling pathway was identified as crucial mediator of cardiomyocyte hypertrophy. In this study we found that activation of non-canonical Wnt signaling by Wnt5a stimulates protein synthesis and enlargement of cardiomyocyte surface area. These hypertrophic features were inhibited in Dapper-1 (Dpr1) depleted cells. On the molecular level, we observed inhibition of the non-canonical Wnt/planar-cell-polarity (PCP) pathway denoted by reduction of c-jun-n-terminal-kinase (JNK) phosphorylation. Upstream of JNK, increased protein levels of the Wnt/PCP trans-membrane receptor van-Gogh-like-2 (Vangl2) were observed along with an enrichment of Vangl2 in perinuclear located vesicles. The findings suggest that Dpr1 is essential for execution of the Wnt/PCP pathway and regulation of the Vangl2/JNK axis. Depletion of Dpr1 inhibits non-canonical Wnt signaling induced cardiomyocyte hypertrophy by blocking Wnt/PCP signaling.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, 10%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00