MilliporeSigma
  • Intestinal microbiota-dependent phosphatidylcholine metabolites, diastolic dysfunction, and adverse clinical outcomes in chronic systolic heart failure.
Intestinal microbiota-dependent phosphatidylcholine metabolites, diastolic dysfunction, and adverse clinical outcomes in chronic systolic heart failure.
Journal of cardiac failure (2014-12-03)
W H Wilson Tang, Zeneng Wang, Kevin Shrestha, Allen G Borowski, Yuping Wu, Richard W Troughton, Allan L Klein, Stanley L Hazen
ABSTRACT
Trimethylamine-N-oxide (TMAO) has been linked to increased cardiovascular risk. We aimed to determine the prognostic value of TMAO and its dietary precursors, choline and betaine, in heart failure (HF). In 112 patients with chronic systolic HF with comprehensive echocardiographic evaluation, we measured plasma TMAO, choline, and betaine by mass spectrometry. Median (interquartile range) TMAO levels, choline, and betaine levels were 5.8 (3.6-12.1) μmol/L, 10.9 (8.4-14.0) μmol/L, and 43.8 (37.1-53.0) μmol/L, respectively, and were correlated with each other (all P < .0001 for both). TMAO levels were significantly higher in patients with diabetes mellitus (9.4 [4.9-13.2] vs 4.8 [3.4-9.8] μmol/L; P = .005) and in subjects with New York Heart Association functional class III or greater (7.0 [4.7-14.8] vs 4.7 [3.4-11.3] μmol/L; P = .02). Elevated TMAO, choline, and betaine levels were each associated with higher plasma N-terminal pro-B-type natriuretic peptide (NT-proBNP) levels and more advanced left ventricular diastolic dysfunction, but not systolic dysfunction or inflammatory and endothelial biomarkers. Higher choline (hazard ratio [HR] 1.64, 95% CI 1.22-2.20; P = .001), betaine (HR 1.51, 95% CI 1.10-2.08; P = .01), and TMAO (HR 1.48, 95% CI 1.10-1.96; P = .01) predicted increased risk for 5-year adverse clinical events (death/transplantation). Only higher TMAO levels predicted incident adverse clinical events independently from age, estimated glomerular filtration rate, mitral E/septal Ea, and NT-proBNP levels (HR 1.46, 95% CI 1.03-2.14; P = .03). Elevated plasma TMAO, choline, and betaine levels are each associated with more advanced left ventricular diastolic dysfunction and portend poorer long-term adverse clinical outcomes in chronic systolic HF. However, only higher plasma TMAO was associated with poor prognosis after adjustment for cardiorenal indices.
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Citrulline, ≥98% (TLC)
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Arginine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)