MilliporeSigma
  • Co-exposure with fullerene may strengthen health effects of organic industrial chemicals.
Co-exposure with fullerene may strengthen health effects of organic industrial chemicals.
PloS one (2014-12-05)
Maili Lehto, Topi Karilainen, Tomasz Róg, Oana Cramariuc, Esa Vanhala, Jarkko Tornaeus, Helena Taberman, Janne Jänis, Harri Alenius, Ilpo Vattulainen, Olli Laine
ABSTRACT
In vitro toxicological studies together with atomistic molecular dynamics simulations show that occupational co-exposure with C60 fullerene may strengthen the health effects of organic industrial chemicals. The chemicals studied are acetophenone, benzaldehyde, benzyl alcohol, m-cresol, and toluene which can be used with fullerene as reagents or solvents in industrial processes. Potential co-exposure scenarios include a fullerene dust and organic chemical vapor, or a fullerene solution aerosolized in workplace air. Unfiltered and filtered mixtures of C60 and organic chemicals represent different co-exposure scenarios in in vitro studies where acute cytotoxicity and immunotoxicity of C60 and organic chemicals are tested together and alone by using human THP-1-derived macrophages. Statistically significant co-effects are observed for an unfiltered mixture of benzaldehyde and C60 that is more cytotoxic than benzaldehyde alone, and for a filtered mixture of m-cresol and C60 that is slightly less cytotoxic than m-cresol. Hydrophobicity of chemicals correlates with co-effects when secretion of pro-inflammatory cytokines IL-1β and TNF-α is considered. Complementary atomistic molecular dynamics simulations reveal that C60 co-aggregates with all chemicals in aqueous environment. Stable aggregates have a fullerene-rich core and a chemical-rich surface layer, and while essentially all C60 molecules aggregate together, a portion of organic molecules remains in water.
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Benzaldehyde, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
SAFC
HEPES
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Acetophenone, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
SAFC
HEPES
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Acetophenone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Toluene, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Benzyl alcohol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
m-Cresol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 g
Estimated to ship on March 24, 2025
$333.00
2.5 kg
Estimated to ship on March 24, 2025
$1,670.00