MilliporeSigma
  • In vitro antioxidant and antitumor activities of six selected plants used in the Traditional Arabic Palestinian herbal medicine.
In vitro antioxidant and antitumor activities of six selected plants used in the Traditional Arabic Palestinian herbal medicine.
Pharmaceutical biology (2014-05-28)
Ahmad Ibrahim Husein, Mohammed Saleem Ali-Shtayeh, Waheed Jebril Jondi, Nidal Abd-Aljapar Zatar, Ibrahim M Abu-Reidah, Rana M Jamous
ABSTRACT
Despite several pharmacological applications of the medicinal plants in the Traditional Arabic Palestinian Herbal Medicine in Palestine (TAPHM), studies on their antioxidant properties are still scarce. This work evaluates the antioxidant and antitumor activities of the ethanol extracts from different parts of six plants: [Arum palaestinum Boiss (Araceae), Urtica pilulifera L. (Urticaceae), Coridothymus capitatus (L.) Reichb (Lamiaceae), Majorana syriaca (L.) Rafin. (Lamiaceae), Teucrium creticum L. (Lamiaceae), and Teucrium capitatum L. (Lamiaceae)] used in the TAPHM. The antioxidant activity was evaluated for the ethanol extracts by DPPH and β-carotene-linoleic acid assays together with total contents of phenols and flavonoids. For the anti-carcinogenic evaluation, the extracts were tested for the ability to inhibit the proliferation of breast cancer cells (MCF-7) using the MTT reduction assay. Among the extracts, the U. pilulifera had the highest amount of total phenolics, possessing the second highest total flavonoids. It also showed a maximum cytotoxic activity (IC50 = 63 µg/ml), followed by C. capitatus, and A. palaestinum. Otherwise, the extract of T. creticum was demonstrated to be an efficient scavenger of O2 (IC50 = 83 µg/ml), followed by M. syriaca, C. capitatus, T. capitatum, A. palaestinum, and U. pilulifera. The results suggest that the investigated plants have shown varied antioxidant capacities which were strongly correlated with their contents of phenolics. Accordingly, this study proposes that the therapeutic benefit of these plants can be, at least in part, attributed to its potential inhibition of oxidative processes.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Guaiacol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
o-Cresol, SAJ first grade, ≥97.0%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
o-Cresol, ≥99%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Supelco
o-Cresol, analytical standard
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%