MilliporeSigma
  • Redox and pH-responsive poly (amidoamine) dendrimer-poly (ethylene glycol) conjugates with disulfide linkages for efficient intracellular drug release.
Redox and pH-responsive poly (amidoamine) dendrimer-poly (ethylene glycol) conjugates with disulfide linkages for efficient intracellular drug release.
Colloids and surfaces. B, Biointerfaces (2014-10-04)
Wen Hu, Liang Cheng, Lifang Cheng, Meng Zheng, Qifu Lei, Ziyang Hu, Ming Xu, Lipeng Qiu, Dawei Chen
ABSTRACT
Nanocarriers with low toxicity, high stability, long circulation in blood and triggered drug release at target sites are extremely needed for cancer therapy. In this study, different molar ratios of poly (ethylene glycol) (PEG) were conjugated to poly (amidoamine) dendrimer (PAMAM) by cleavable disulfide bonds, yielding redox and pH dual-responsive nanocarriers (PSSP). The redox sensitivity of the PSSP conjugates was confirmed by measuring the change in particle size and zeta potential under reductive environment by dynamic light scattering (DLS). Meanwhile, all conjugates showed slight hemolytic ratio and high stability in 10% FBS for 24h. Phagocytosis experiments indicated that the PSSP conjugates could effectively escape the engulfment of macrophage. Doxorubicin (DOX) was loaded into the hydrophobic core of the conjugates with loading content of 10%. In vitro release studies suggested that release rate as well as cumulative release amount of DOX from the PSSP conjugates were pronouncedly enhanced in reductive environment as compared with the non-cleavable counterparts. Besides, all conjugates exhibited excellent acid-triggered release performance. Significantly, PEGylation degree of the conjugates had critical impact on drug release. Cytotoxicity tests displayed that DOX-loaded PSSP conjugates were more potent in killing B16 and A549 tumor cells than the non-cleavable counterparts. Curiously, PSSP conjugate with the highest PEGylation degree showed the best antitumor effect despite the somewhat less cell uptake. Therefore, the dual-responsive PSSP conjugates seem promising candidates for efficient intracellular release of antitumor drugs.
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Supelco
Triethylamine, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, NMR reference standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, SAJ special grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Estimated to ship on March 23, 2025
$199.00
25 g
Estimated to ship on March 23, 2025
$628.00
100 g
Estimated to ship on March 23, 2025
$2,160.00
500 g
Estimated to ship on March 23, 2025
$3,580.00