MilliporeSigma
  • Determinants for achieving the LDL-C target of lipid control for secondary prevention of cardiovascular events in Taiwan.
Determinants for achieving the LDL-C target of lipid control for secondary prevention of cardiovascular events in Taiwan.
PloS one (2015-03-11)
Li-Ting Ho, Wei-Hsian Yin, Shao-Yuan Chuang, Wei-Kung Tseng, Yen-Wen Wu, I-Chang Hsieh, Tsung-Hsien Lin, Yi-Heng Li, Lien-Chi Huang, Kuo-Yang Wang, Kwo-Chang Ueng, Ching-Chang Fang, Wen-Harn Pan, Hung-I Yeh, Chau-Chung Wu, Jaw-Wen Chen
ABSTRACT
Epidemiological and clinical studies have clearly established the link between low-density lipoprotein cholesterol (LDL-C) and atherosclerosis-related cardiovascular consequences. Although it has been a common practice for physicians to prescribe lipid-lowering therapy for patients with dyslipidemia, the achievement rate is still not satisfied in Taiwan. Therefore, the determinants for achieving the LDL-C target needed to be clarified for better healthcare of the patients with dyslipidemia. This registry-type prospective observational study enrolled the patients with cardiovascular diseases (coronary artery disease (CAD) and cerebrovascular disease (CVD)) from 18 medical centers across Taiwan, and clinically followed them for five years. At every clinical visit, vital signs, clinical endpoints, adverse events, concurrent medications and laboratory specimens were obtained as thoroughly as possible. The lipid profile (total cholesterol, high-density lipoprotein cholesterol, LDL-C, triglyceride), liver enzymes, and creatinine phosphokinase were evaluated at baseline, and every year thereafter. The cross sectional observational data was analyzed for this report. Among the 3,486 registered patients, 54% had their LDL-C < 100 mg/dL. By univariate analysis, the patients achieving the LDL-C target were associated with older age, more male sex, taller height, lower blood pressure, more under lipid-lowering therapy, more smoking cessation, more history of CAD, DM, physical activity, but less history of CVD. The multivariate analysis showed statin therapy was the most significant independent determinant for achieving the treatment target, followed by age, history of CAD, diabetes, blood pressure, and sex. However, most patients were on regimens of very-low to low equipotent doses of statins. Although the lipid treatment guideline adherence is improving in recent years, only 54% of the patients with cardiovascular diseases have achieved their LDL-C target in Taiwan, and the most significant determinant for this was statin therapy.
Product Description
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Dibutyl phthalate, SAJ special grade, ≥98.0%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, SAJ first grade, ≥98.0
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)