MilliporeSigma
  • Selective protection of the cerebellum against intracerebroventricular LPS is mediated by local melatonin synthesis.
Selective protection of the cerebellum against intracerebroventricular LPS is mediated by local melatonin synthesis.
Brain structure & function (2013-12-24)
Luciana Pinato, Sanseray da Silveira Cruz-Machado, Daiane G Franco, Leila M G Campos, Erika Cecon, Pedro A C M Fernandes, Jackson C Bittencourt, Regina P Markus
ABSTRACT
Although melatonin is mainly produced by the pineal gland, an increasing number of extra-pineal sites of melatonin synthesis have been described. We previously demonstrated the existence of bidirectional communication between the pineal gland and the immune system that drives a switch in melatonin production from the pineal gland to peripheral organs during the mounting of an innate immune response. In the present study, we show that acute neuroinflammation induced by lipopolysaccharide (LPS) injected directly into the lateral ventricles of adult rats reduces the nocturnal peak of melatonin in the plasma and induces its synthesis in the cerebellum, though not in the cortex or hippocampus. This increase in cerebellar melatonin content requires the activation of nuclear factor kappa B (NF-κB), which positively regulates the expression of the key enzyme for melatonin synthesis, arylalkylamine N-acetyltransferase (AA-NAT). Interestingly, LPS treatment led to neuronal death in the hippocampus and cortex, but not in the cerebellum. This privileged protection of cerebellar cells was abrogated when G-protein-coupled melatonin receptors were blocked by the melatonin antagonist luzindole, suggesting that the local production of melatonin protects cerebellar neurons from LPS toxicity. This is the first demonstration of a switch between pineal and extra-pineal melatonin production in the central nervous system following a neuroinflammatory response. These results have direct implications concerning the differential susceptibility of specific brain areas to neuronal death.
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Luzindole, ≥90%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)