MilliporeSigma
  • High-throughput analysis of gene essentiality and sporulation in Clostridium difficile.
High-throughput analysis of gene essentiality and sporulation in Clostridium difficile.
mBio (2015-02-26)
Marcin Dembek, Lars Barquist, Christine J Boinett, Amy K Cain, Matthew Mayho, Trevor D Lawley, Neil F Fairweather, Robert P Fagan
ABSTRACT
Clostridium difficile is the most common cause of antibiotic-associated intestinal infections and a significant cause of morbidity and mortality. Infection with C. difficile requires disruption of the intestinal microbiota, most commonly by antibiotic usage. Therapeutic intervention largely relies on a small number of broad-spectrum antibiotics, which further exacerbate intestinal dysbiosis and leave the patient acutely sensitive to reinfection. Development of novel targeted therapeutic interventions will require a detailed knowledge of essential cellular processes, which represent attractive targets, and species-specific processes, such as bacterial sporulation. Our knowledge of the genetic basis of C. difficile infection has been hampered by a lack of genetic tools, although recent developments have made some headway in addressing this limitation. Here we describe the development of a method for rapidly generating large numbers of transposon mutants in clinically important strains of C. difficile. We validated our transposon mutagenesis approach in a model strain of C. difficile and then generated a comprehensive transposon library in the highly virulent epidemic strain R20291 (027/BI/NAP1) containing more than 70,000 unique mutants. Using transposon-directed insertion site sequencing (TraDIS), we have identified a core set of 404 essential genes, required for growth in vitro. We then applied this technique to the process of sporulation, an absolute requirement for C. difficile transmission and pathogenesis, identifying 798 genes that are likely to impact spore production. The data generated in this study will form a valuable resource for the community and inform future research on this important human pathogen. Clostridium difficile is a common cause of potentially fatal intestinal infections in hospital patients, particularly those who have been treated with antibiotics. Our knowledge of this bacterium has been hampered by a lack of tools for dissecting the organism. We have developed a method to study the function of every gene in the bacterium simultaneously. Using this tool, we have identified a set of 404 genes that are required for growth of the bacteria in the laboratory. C. difficile also produces a highly resistant spore that can survive in the environment for a long time and is a requirement for transmission of the bacteria between patients. We have applied our genetic tool to identify all of the genes required for production of a spore. All of these genes represent attractive targets for new drugs to treat infection.
Sigma-Aldrich
3-Methyl-1-butanol, JIS special grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Methyl-1-butanol, SAJ first grade, ≥96.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, BioPerformance Certified
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
E-Toxate Water, endotoxin, free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for molecular biology, sterile filtered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Methylbutanol, for molecular biology, BioReagent, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG