Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium Fluoride Solution
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, Vetec™, reagent grade, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sign Into View Organizational & Contract Pricing