MilliporeSigma
  • Real-time monitoring of the metabolic activity of periodontopathic bacteria.
Real-time monitoring of the metabolic activity of periodontopathic bacteria.
Journal of microbiological methods (2015-05-20)
Kazuko Ishiguro, Jumpei Washio, Keiichi Sasaki, Nobuhiro Takahashi
ABSTRACT
Bacterial metabolic activity is associated with the onset and progression mechanisms of oral biofilm-mediated disease; however, at present no method to monitor bacterial metabolism exists, especially for periodontopathic bacteria. Therefore, we aimed to establish a novel method for monitoring the metabolic activity of periodontopathic bacteria, Porphyromonas gingivalis (Pg), Prevotella intermedia (Pi) and Fusobacterium nucleatum (Fn), as well as Streptococcus mutans (Sm) for comparison. The method is based on the dye resazurin, which is converted to the fluorescent molecule resorufin by reducing molecules derived from bacterial metabolism. Additionally, the effects of antimicrobial substances on bacterial metabolic activity were evaluated using this method. When bacterial suspensions were incubated with tryptone, glutamate, aspartate or glucose in the presence of resazurin, the fluorescence intensity increased over time by these bacterial metabolic reactions, indicating that this method can be used to monitor the metabolic activity of periodontopathic bacteria. Chlorhexidine showed the 50% inhibitory concentration (IC50) of 15-49 μg/ml for tryptone metabolism by Pg, Pi, and Fn, and 7.1-18 μg/ml for glucose metabolism by Pi and Sm. The IC50s for cetylpyridinium chloride and sodium dodecyl sulfate were 0.8-2.1 and 28-44 μg/ml, respectively for all bacteria examined. Fluoride had no effect except the IC50 of 640 μg/ml for Sm, while minocycline hydrochloride had no effect on any of the bacteria. The present study established the method for real-time monitoring of the metabolic activity of periodontopathic bacteria, and the method might be useful for evaluating the effects of antimicrobial substances on the bacterial metabolic activity.
Sigma-Aldrich
Resorufin, 95%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hexadecylpyridinium chloride monohydrate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Minocycline hydrochloride, powder
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture