MilliporeSigma
  • Biotransformation of luteoloside by a newly isolated human intestinal bacterium using UHPLC-Q-TOF/MS.
Biotransformation of luteoloside by a newly isolated human intestinal bacterium using UHPLC-Q-TOF/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-04-23)
Jin-hua Tao, Dong-geng Wang, Chi Yang, Jin-hua Huang, Wen-qian Qiu, Xi Zhao
ABSTRACT
To explore the metabolic pathways and metabolites of luteoloside yielded by the isolated human intestinal bacteria from healthy human feces and characterize the β-d-glucosidase activity of the specific strain which catalyzed the breakdown of luteoloside, a preculture bacterial GAM broth and luteoloside were mixed incubated together for 48h. UHPLC-Q-TOF/MS was used for analysis of the metabolites of luteoloside in the corresponding supernatant fractions from fermentation. Aliquots of the reactive solutions were collected at different times and were measured with a microplate reader at 405nm to evaluate the enzymatic activity. Three metabolites (acetylated luteoloside, luteolin and deoxygenated luteolin) were detected in the fractions isolated from the bacterial samples. The variation of β-d-glucosidase activity inside the bacterium was in coincidence with the changes in luteolin generation or luteoloside degradation in different time periods.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%