MilliporeSigma
  • A rapid method for chemical fingerprint analysis of Pan Panax notoginseng powders by ultra performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry.
A rapid method for chemical fingerprint analysis of Pan Panax notoginseng powders by ultra performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry.
Chinese journal of natural medicines (2015-06-16)
Peng Liu, He-Shuil Yu, Li-Juan Zhang, Xin-Bo Song, Li-Ping Kang, Jing-Yuan Liu, Jie Zhang, Man Cao, Kate Yu, Ting-Guo Kang, Bai-Ping Ma
ABSTRACT
A method coupling ultra-performance liquid chromatography (UPLC) with quadrupole time-of-flight mass spectrometer (Qtof MS) using the electrospray ionization (ESI) source was developed for the identification of the major saponins from Panax notoginseng powder (PNP). Ten different PNP samples were analyzed and evaluated for their quality by similarity evaluation and principle component analysis (PCA). Based on the accurate mass, summarized characteristic fragmentation behaviors, retention times of different types of saponins, related botanical biogenesis, and reported chromatographic behavior of saponins, fifty-one common peaks were effectively separated and identified, including 28 protopanaxadiol saponins and 18 protopanaxatriol saponins. Simultaneously, 15 significant discrepancy compounds were identified from the disqualified PNP samples. The established UPLC/Qtof MS fingerprint method was successfully applied for profiling and identifying the major saponins of PNP, providing a fast quality evaluation tool for distinguishing the authentic PNP and the adulterated products.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG