MilliporeSigma
  • Interaction of osmoregulatory and acid-base compensation in white sturgeon (Acipenser transmontanus) during exposure to aquatic hypercarbia and elevated salinity.
Interaction of osmoregulatory and acid-base compensation in white sturgeon (Acipenser transmontanus) during exposure to aquatic hypercarbia and elevated salinity.
The Journal of experimental biology (2015-09-04)
Ciaran A Shaughnessy, Dan W Baker, Colin J Brauner, John D Morgan, Jason S Bystriansky
ABSTRACT
Migratory fishes encounter a variety of environmental conditions, including changes in salinity, temperature and dissolved gases, and it is important to understand how these fishes are able to acclimate to multiple environmental stressors. The gill is the primary site of both acid-base balance and ion regulation in fishes. Many ion transport mechanisms involved with acid-base compensation are also required for the regulation of plasma Na(+) and Cl(+), the predominant extracellular ions, potentially resulting in a strong interaction between ionoregulation and acid-base regulation. The present study examined the physiological interaction of elevated dissolved CO2 (an acid-base disturbance) on osmoregulation during seawater acclimation (an ionoregulatory disturbance) in juvenile white sturgeon (Acipenser transmontanus). Blood pH (pHe), plasma [HCO3 (-)], [Na(+)], [Cl(-)] and osmolality, white muscle water content, and gill Na(+)/K(+)-ATPase (NKA) and Na(+)/K(+)/2Cl(-) co-transporter (NKCC) abundance were examined over a 10 day seawater (SW) acclimation period under normocarbia (NCSW) or during prior and continued exposure to hypercarbia (HCSW), and compared with a normocarbic freshwater (NCFW) control. Hypercarbia induced a severe extracellular acidosis (from pH 7.65 to pH 7.2) in HCSW sturgeon, and these fish had a 2-fold greater rise in plasma osmolarity over NCSW by day 2 of SW exposure. Interestingly, pHe recovery in HCSW was associated more prominently with an elevation in plasma Na(+) prior to osmotic recovery and more prominently with a reduction in plasma Cl(-) following osmotic recovery, indicating a biphasic response as the requirements of osmoregulation transitioned from ion-uptake to ion-excretion throughout SW acclimation. These results imply a prioritization of osmoregulatory recovery over acid-base recovery in this period of combined exposure to acid-base and ionoregulatory disturbances.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, tablet
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride solution, 1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
SAFC
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00