MilliporeSigma
  • Anthocyanin Profile and Antioxidant Activity of Various Berries Cultivated in Korea.
Anthocyanin Profile and Antioxidant Activity of Various Berries Cultivated in Korea.
Natural product communications (2015-07-23)
Hong-Sook Bae, Hyun Ju Kim, Jin Hee Kang, Rika Kudo, Takahiro Hosoya, Shigenori Kumazawa, Mira Jun, Oh-Yoen Kim, Mok-Ryeon Ahn
ABSTRACT
This study examined the anthocyanin composition and antioxidant activity of various berries cultivated in Korea: blueberry, crowberry, Korean black raspberry, mulberry, and strawberry. The anthocyanins in berries were identified by high-performance liquid chromatography (HPLC) analysis, and each component was quantitatively analyzed. Furthermore, the antioxidant activity of berries was evaluated using 2,2-diphenyl-1-picrylhydrazyl (DPPH) free radical-scavenging, 2,2'-azinobis (3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) radical cation decolorization, oxygen radical absorbance capacity (ORAC), and ferric ion reducing antioxidant (FRAP) assays. The results revealed that the total content of anthocyanins in crowberry was 35.1 mg/g of extract, which was higher than that in the other four major berry species (1.9-27.7 mg/g of extract). Nineteen anthocyanins were identified in the various berries. The major anthocyanins of crowberry were cyanidin-3-galactoside and delphinidin-3-galactoside, and those from Korean black raspberry were cyanidin-3-rutinoside and cyanidin-3-sambubioside-5-rhamnoside. These two berries also had relatively strong antioxidant activity accompanied by high total polyphenol contents. Thus, consumption of crowberry and Korean black raspberry may be beneficial in reducing the risk of developing lifestyle-related chronic diseases because of their strong antioxidant activity.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Aluminum chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles