MilliporeSigma
  • Imidazole-Modified Nanoporous Silica for Lead Ion Solid Phase Extraction Prior to Determination from Industrial Wastewaters by Flame Atomic Absorption Spectrometry.
Imidazole-Modified Nanoporous Silica for Lead Ion Solid Phase Extraction Prior to Determination from Industrial Wastewaters by Flame Atomic Absorption Spectrometry.
Journal of AOAC International (2015-11-04)
Ali Behbahani, Mehdi Ardjmand
ABSTRACT
A new method was applied to produce modified nanoporous silica as a novel sorbent for Pb(II) ion SPE from industrial wastewater samples. In this modified method, the produced nanoporous silica has a higher functional group loading, which leads to a higher preconcentration factor as well as a lower LOD. This modified nanoporous silica was used for preconcentration prior to subsequent determination of Pb(II) ions by flame atomic absorption spectrometry. Various parameters such as the eluent, pH of the sample solution, and flow rate were optimized during this work. Also, the effect of a variety of ions on preconcentration and recovery of Pb(II) ions was investigated. The LOD, defined as five times the SD of the blank, was determined to be lower than 0.1 mg/L with an RSD of <2%. The accuracy of the method was established by analyzing standard reference materials with certified Pb concentrations. Finally, the established method was successfully applied for determination of the Pb(II) ion concentration in industrial wastewater samples.
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxalyl chloride, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxalyl chloride, reagent grade, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethylamine, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxalyl chloride solution, 2.0 M in methylene chloride
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%