MilliporeSigma
  • DNA-PKcs plays role in cancer metastasis through regulation of secreted proteins involved in migration and invasion.
DNA-PKcs plays role in cancer metastasis through regulation of secreted proteins involved in migration and invasion.
Cell cycle (Georgetown, Tex.) (2015-05-29)
Ewa Kotula, Nathalie Berthault, Celine Agrario, Marie-Christine Lienafa, Anthony Simon, Florent Dingli, Damarys Loew, Vonick Sibut, Simon Saule, Marie Dutreix
ABSTRACT
The DNA-dependent protein kinase catalytic subunit (DNA-PKcs) plays a major role in DNA damage signaling and repair and is also frequently overexpressed in tumor metastasis. We used isogenic cell lines expressing different levels of DNA-PKcs to investigate the role of DNA-PKcs in metastatic development. We found that DNA-PKcs participates in melanoma primary tumor and metastasis development by stimulating angiogenesis, migration and invasion. Comparison of conditioned medium content from DNA-PKcs-proficient and deficient cells reveals that DNA-PKcs controls secretion of at least 103 proteins (including 44 metastasis-associated with FBLN1, SERPINA3, MMP-8, HSPG2 and the inhibitors of matrix metalloproteinases, such as α-2M and TIMP-2). High throughput analysis of secretomes, proteomes and transcriptomes, indicate that DNA-PKcs regulates the secretion of 85 proteins without affecting their gene expression. Our data demonstrate that DNA-PKcs has a pro-metastatic activity via the modification of the tumor microenvironment. This study shows for the first time a direct link between DNA damage repair and cancer metastasis and highlights the importance of DNA-PKcs as a potential target for anti-metastatic treatment.
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
NU7026, ≥98% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium trichloroacetate, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKDC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Prkdc
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), colorimetric, The CHEMICON Cell Invasion Assay Kit uses a 24-well plate, with 8 um pores, which provides an efficient system for evaluating the invasion of tumor cells through a basement membrane model.