MilliporeSigma
  • Residue Depletion Study of Danofloxacin in Cultured Tilapia (Oreochromis mossambicus).
Residue Depletion Study of Danofloxacin in Cultured Tilapia (Oreochromis mossambicus).
Journal of AOAC International (2015-05-31)
Yang-Chi Fan, Shi-Yuan Sheu, Hong-Thih Lai, Ming-Huang Chang, Pei-Heng Chen, Yi-Chih Lei, Tzong-Fu Kuo, Chia-Yih Wang
ABSTRACT
Danofloxacin is an antibacterial drug of the fluoroquinolone group developed for therapeutic purposes in veterinary medicine. The studies described here include investigations of the residues following a single dose or multiple doses of danofloxacin. Residue depletion studies were performed to determine residues in plasma and tissues of saltwater tilapia fish (Oreochromis mossambicus) after a single oral administration of danofloxacin at the dose of 10 mg/kg body weight and also after daily dose of 10 mg/kg body weight for five consecutive days. Danofloxacin residues were analyzed by HPLC with fluorescence detection. Following a single oral dose, danofloxacin residues in 6 h postdosing tilapia were at a maximum of 1.44, 12.48, and 13.18 μg/g in serum, liver, and kidney samples, respectively, while a peak muscle concentration of 2.15 μg/g was reached at 12 h. From single-dose data, the concentration of danofloxacin in serum, muscle, liver, and kidney samples declined with half-lives of 29, 34, 49, and 44 h, respectively. Based on the maximum residue level (MRL) of 0.1 μg/g in edible tissue for fin fish, the withdrawal times of danofloxacin in muscle were estimated to fall below the MRL after a withdrawal period of 21 days following the multiple-dose administration. These results may be helpful to regulatory agencies as they determine what tissues should be monitored to ensure that the established residue safety tolerance levels are not exceeded.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol