MilliporeSigma
  • An LC-MS/MS method for simultaneous determination of three Polygala saponin hydrolysates in rat plasma and its application to a pharmacokinetic study.
An LC-MS/MS method for simultaneous determination of three Polygala saponin hydrolysates in rat plasma and its application to a pharmacokinetic study.
Journal of ethnopharmacology (2015-04-30)
Qian Wang, Bing-Xin Xiao, Rui-Le Pan, Xin-Min Liu, Yong-Hong Liao, Li Feng, Fang-Rui Cao, Qi Chang
ABSTRACT
Radix Polygala has a long history of use as a sedative in traditional Chinese medicine and its major ingredients are saponins, which are recognized effective in memory improvement but highly toxic to gastricintestinal mucosa. Polygala saponin hydrolysates (PSH), an alkaline hydrolysis product and also the intestinal metabolites of the saponins, exhibited stronger effects in improving memory of mice and had less toxicity than its original saponins. The present study aims to develop a sensitive LC-MS/MS method for simultaneously determining PSH three major active components, 3,4,5-trimethoxycinnamylic acid (TMCA), p-methoxycinnamylic acid (PMCA) and tenuifolin (TF), in rat plasma and apply the method to a pharmacokinetic study. The acidic plasma (100μl) was treated by liquid-liquid extraction with ethyl acetate and reconstituted sample was analyzed on a C18 column eluted with acetonitrile-water (50:50) containing 0.2% formic acid at 0.4ml/min. The mass detection in negative electrospray ionization was used. The ion pairs for multiple reaction monitoring were set at m/z 237.0/103.0, 177.0/116.6 and 679.5/425.3 for TMCA, PMCA and TF, respectively. Their pharmacokinetic profiles were studied in rats after intravenous and oral dose of PSH at 20 and 100mg/kg, respectively. The calibration curves had good linearity (r(2)>0.99) for TMCA, PMCA and TF within the tested concentration ranges. The limits of detection and quantification were 1, 10, 0.5ng/ml and 10.0, 20.0, 1.0ng/ml, respectively. The intra-day and inter-day precisions were less than 18.9% and accuracies between 93.2% and 113.3%, and the extraction recovery ranged from 91.2% to 112.1% for all analytes. The pharmacokinetic study showed that TMCA, PMCA and TF could be rapidly absorbed into the circulation and reached their peak concentrations at about 9.1, 9.0 and 24.0min, respectively. TF had a lower oral bioavailability (2.0%) than TMCA (90.1%) and PMCA (96.5%), but it remained in the body much longer (t1/2, λz, 4.8h, oral dose) than TMCA (0.6h) and PMCA (0.9h). A sensitive LC-MS/MS method was developed and applied to a pharmacokinetic study of TMCA, PMCA and TF of PSH in rats. The three components are proved to be bio-available active components of PSH and might display their in vivo pharmacological activities at different levels and different time periods after oral administration.
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥70% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%