MilliporeSigma
Effect of antioxidants on the genotoxicity of phenethyl isothiocyanate.
Mutagenesis (2015-02-15)
Jared D Hoffman, William M Ward, George Loo
ABSTRACT
Isothiocyanates are plant-derived compounds that may be beneficial in the prevention of certain chronic diseases. Yet, by stimulating the production of reactive oxygen species (ROS), isothiocyanates can be genotoxic. Whether antioxidants influence isothiocyanate-induced genotoxicity is unclear, but this situation was clarified appreciably herein. In HCT116 cells, phenethyl isothiocyanate (PEITC) increased ROS production, which was inhibited by N-acetylcysteine (NAC) and deferoxamine (DFO) but not by ascorbic acid (ASC) and trolox (TRX) that were found to be more potent radical scavengers. Surprisingly, ASC and TRX each intensified the DNA damage that was caused by PEITC, but neither ASC nor TRX by themselves caused any DNA damage. In contrast, NAC and DFO each not only attenuated PEITC-induced DNA damage but also attenuated the antioxidant-intensified, PEITC-induced DNA damage. To determine if the DNA damage could be related to possible changes in the major antioxidant defence system, glutathione (GSH) was investigated. PEITC lowered GSH levels, which was prevented by NAC, whereas ASC, TRX and DFO neither inhibited nor enhanced the GSH-lowering effect of PEITC. The GSH synthesis inhibitor, buthionine sulphoxime, intensified PEITC-induced DNA damage, although by itself buthionine sulphoxime did not directly cause DNA damage. The principal findings suggest that ASC and TRX make PEITC more genotoxic, which might be exploited in killing cancer cells as one approach in killing cancer cells is to extensively damage their DNA so as to initiate apoptosis.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, reagent grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,2′-Dipyridyl, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Tocopherol, ≥95.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Acetyl-L-cysteine, SAJ special grade, 98.0-102.0%