MilliporeSigma
  • Development of a quantitative method for the simultaneous analysis of the boar taint compounds androstenone, skatole and indole in porcine serum and plasma by means of ultra-high performance liquid chromatography coupled to high resolution mass spectrometry.
Development of a quantitative method for the simultaneous analysis of the boar taint compounds androstenone, skatole and indole in porcine serum and plasma by means of ultra-high performance liquid chromatography coupled to high resolution mass spectrometry.
Food chemistry (2015-05-16)
J Wauters, J Vanden Bussche, K Verplanken, K M Bekaert, M Aluwé, A Van den Broeke, A Coussé, N Buys, L Vanhaecke
ABSTRACT
Boar taint is an off-odour occurring while heating meat or fat from boars. A method detecting the three compounds (androstenone, skatole and indole) simultaneously in blood would offer substantial advantages since it would allow monitoring the impact of rearing strategies. Therefore, a UHPLC-HR-Orbitrap-MS analysis method is optimized and validated for the quantification of these compounds in plasma or serum. Sample pre-treatment involved an extraction with diethylether followed by a centrifugal filtration (30 kDa). Limits of detection and quantification varied between 0.5 and 1 μg L(-1) and 2 and 3 μg L(-1) for the three compounds, respectively. Besides, an excellent repeatability (RSD < 7.6%), within-laboratory reproducibility (RSD<10.5%), recovery (87-97%) and linearity (R(2)>0.99) were recorded. Correlations between serum/plasma and fat levels of the boar taint compounds were positive for skatole (r(serum) = 0.39 and r(plasma) = 0.84) and androstenone (r(serum) = 0.73-0.78 and r(plasma) = 0.32-0.80).
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Skatole, ≥98%
Sigma-Aldrich
5α-Androst-16-en-3-one
Sigma-Aldrich
2-Methylindole, 98%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
1,4-Androstadiene-3,17-dione
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
3-Methylindole, 98%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300