MilliporeSigma
  • An optical biosensor assay for rapid dual detection of Botulinum neurotoxins A and E.
An optical biosensor assay for rapid dual detection of Botulinum neurotoxins A and E.
Scientific reports (2015-12-10)
Christian Lévêque, Géraldine Ferracci, Yves Maulet, Christelle Mazuet, Michel R Popoff, Marie-Pierre Blanchard, Michael Seagar, Oussama El Far
ABSTRACT
The enzymatic activity of the pathogenic botulinum neurotoxins type A and E (BoNT/A and E) leads to potentially lethal paralytic symptoms in humans and their prompt detection is of crucial importance. A chip assay based on Surface Plasmon Resonance monitoring of the cleavage products is a simple method that we have previously established to detect BoNT/A activity. We have now developed a similar format assay to measure BoNT/E activity. A monoclonal antibody specifically recognizing SNAP25 cleaved by BoNT/E was generated and used to measure the appearance of the neo-epitope following injection of BoNT/E over SNAP-25 immobilized on a chip. This assay detects BoNT/E activity at 1 LD50/ml within minutes and linear dose-responses curves were obtained using a multiplexed biosensor. A threshold of 0.01 LD50/ml was achieved after 5 h of cleavage. This assay is 10-fold more sensitive than the in vivo assay for direct detection of BoNT/E in serum samples. The SNAP25 chip assay is able to discriminate in an automated manner the presence of BoNT/E, BoNT/A or a combination of both toxins.
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%