MilliporeSigma
  • Quantitative assessment of human serum transferrin receptor in breast cancer patients pre- and post-chemotherapy using peptide immunoaffinity enrichment coupled with targeted proteomics.
Quantitative assessment of human serum transferrin receptor in breast cancer patients pre- and post-chemotherapy using peptide immunoaffinity enrichment coupled with targeted proteomics.
Clinica chimica acta; international journal of clinical chemistry (2015-06-23)
Qingqing Xu, Mingchen Zhu, Ting Yang, Feifei Xu, Yuan Liu, Yun Chen
ABSTRACT
sTfR, a soluble form of transferrin receptor in serum, has been suggested as an indicator of bone marrow failure in breast cancer patients receiving chemotherapy. However, intensive chemotherapy could also cause a reduction of sTfR to a level below the LOQ of most assays. An advanced liquid chromatography-tandem mass spectrometry (LC-MS/MS)-based targeted proteomics assay coupled with peptide immunoaffinity enrichment (SISCAPA) was developed and validated for monitoring sTfR. Tryptic peptide 681VEYHFLSPYVSPK693 was selected as a surrogate analyte for quantification. High-abundant proteins were first removed from serum, followed by SISCAPA that was effective in surrogate peptide enrichment and sensitivity enhancement. The resulting LOQ can achieve 100ng/ml (~10-fold increase). Then, sTfR levels in breast cancer patients pre- and post-chemotherapy, and healthy volunteers were accurately quantified as 1.77±0.53μg/ml, 0.98±0.26μg/ml and 1.66±0.50μg/ml, respectively, using a standard addition method. While there was no evidence for a difference between patients and healthy volunteers, differential levels of sTfR pre- and post-chemotherapy were obtained. Comparison between SISCAPA-targeted proteomics and ELISA indicated that the former approach provided a lower value of sTfR. SISCAPA-targeted proteomics may allow the quantification of low-abundant proteins in a more accurate manner.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0%
Sigma-Aldrich
3-Indoleacetic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))